Assay Design



Gene Transcripts

Gene Symbol
ALDH5F1
Entrez Gene ID
844282
Gene Name
aldehyde dehydrogenase 5F1
Gene Aliases
AT1G79440, ENF1, ENLARGED FIL EXPRESSION DOMAIN 1, SSADH, SSADH1, SUCCINIC SEMIALDEHYDE DEHYDROGENASE, SUCCINIC SEMIALDEHYDE DEHYDROGENASE 1, aldehyde dehydrogenase 5F1
Chromosome Location
Chr.1: 29882121 - 29887414 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_106592.5 NP_178062.1 7 - 8 767 62
GenBank mRNA AF117335.1 - 7 - 8 685 62
AF428367.1 - 7 - 8 687 62
AY056147.1 - 7 - 8 673 62

View Full Product Details