Assay Design
Gene Transcripts
- Gene Symbol
- GA2
- Entrez Gene ID
-
844284
- Gene Name
- Terpenoid cyclases/Protein prenyltransferases superfamily protein
- Gene Aliases
-
ARABIDOPSIS THALIANA ENT-KAURENE SYNTHASE, ARABIDOPSIS THALIANA ENT-KAURENE SYNTHASE 1, AT1G79460, ATKS, ATKS1, ENT-KAURENE SYNTHASE, ENT-KAURENE SYNTHASE 1, GA REQUIRING 2, KS, KS1, T8K14.12, T8K14_12
- Chromosome Location
-
Chr.1: 29890367 - 29894646 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_106594.4 |
NP_178064.1 |
14 - 15 |
2141 |
99 |
View Full Product Details