Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001334912.1 | NP_001320544.1 | 3 - 4 | 386 | 84 |
NM_106618.5 | NP_565218.1 | 2 - 3 | 558 | 84 | |
GenBank mRNA | AY120774.1 | - | 2 - 3 | 274 | 84 |
BT000142.1 | - | 2 - 3 | 244 | 84 |