Assay Design



Gene Transcripts

Gene Symbol
WIN1
Entrez Gene ID
844399
Gene Name
HOPW1-1-interacting 1
Gene Aliases
AT1G80600, HOPW1-1-interacting 1, T21F11.7, T21F11_7, TUMOR PRONE 5, TUP5
Chromosome Location
Chr.1: 30298462 - 30300658 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_106708.4 NP_178175.1 1 - 2 560 64
GenBank mRNA AY054594.1 - 1 - 2 455 64
BT002584.1 - 1 - 2 415 64
EU214908.1 - 1 - 2 415 64

View Full Product Details