Assay Design
Gene Transcripts
- Gene Symbol
- ZFP1
- Entrez Gene ID
-
844412
- Gene Name
- zinc-finger protein 1
- Gene Aliases
-
ARABIDOPSIS THALIANA ZINC-FINGER PROTEIN 1, AT1G80730, ATZFP1, F23A5.8, F23A5_8, zinc-finger protein 1
- Chromosome Location
-
Chr.1: 30339322 - 30340385 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_106721.2 |
NP_178188.1 |
1 - 1 |
895 |
149 |
GenBank mRNA |
L39644.1 |
- |
1 - 1 |
757 |
149 |
View Full Product Details