Assay Design



Gene Transcripts

Gene Symbol
WRKY40
Entrez Gene ID
844423
Gene Name
WRKY DNA-binding protein 40
Gene Aliases
AT1G80840, ATWRKY40, F23A5.19, F23A5_19, WRKY DNA-binding protein 40
Chromosome Location
Chr.1: 30383561 - 30385499 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_106732.4 NP_178199.1 2 - 3 702 142
GenBank mRNA AB493542.1 - 2 - 3 429 142
AF480149.1 - 2 - 3 429 142
BT001914.1 - 2 - 3 429 142

View Full Product Details