Assay Design



Gene Transcripts

Gene Symbol
AT3G09100
Entrez Gene ID
820064
Gene Name
mRNA capping enzyme family protein
Gene Aliases
AT3G09100
Chromosome Location
Chr.3: 2787953 - 2793547 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_111745.3 NP_187522.2 5 - 6 804 115
NM_202534.2 NP_974263.1 5 - 6 819 115
GenBank mRNA AY125498.1 - 5 - 6 798 115

View Full Product Details