Assay Design



Gene Transcripts

Gene Symbol
ATB' BETA
Entrez Gene ID
820146
Gene Name
Protein phosphatase 2A regulatory B subunit family protein
Gene Aliases
AT3G09880
Chromosome Location
Chr.3: 3029471 - 3032399 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_111823.3 NP_187599.1 1 - 2 1589 65
GenBank mRNA AY072226.1 - 1 - 2 1549 65
AY113852.1 - 1 - 2 1158 65
U73527.1 - 1 - 2 1526 65

View Full Product Details