Assay Design



Gene Transcripts

Gene Symbol
RLP35
Entrez Gene ID
820279
Gene Name
receptor like protein 35
Gene Aliases
AT3G11080, AtRLP35, F11B9.4, receptor like protein 35
Chromosome Location
Chr.3: 3470304 - 3473685 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001337913.1 NP_001319521.1 1 - 1 1779 98
NM_111945.2 NP_187719.1 1 - 1 1643 98

View Full Product Details