Assay Design



Gene Transcripts

Gene Symbol
ELO1
Entrez Gene ID
820292
Gene Name
Paxneb protein-like protein
Gene Aliases
AT3G11220, ELONGATA 1
Chromosome Location
Chr.3: 3513350 - 3516447 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001337916.1 NP_001319522.1 7 - 8 1164 81
NM_111957.5 NP_566388.2 7 - 8 1066 81

View Full Product Details