Assay Design



Gene Transcripts

Gene Symbol
CARA
Entrez Gene ID
822396
Gene Name
carbamoyl phosphate synthetase A
Gene Aliases
AT3G27740, CARBAMOYL PHOSPHATE SYNTHETASE SMALL SUBUNIT, VEN6, VENOSA 6, carbamoyl phosphate synthetase A
Chromosome Location
Chr.3: 10281230 - 10283935 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_113690.3 NP_566824.1 4 - 5 564 79
GenBank mRNA AY046004.1 - 4 - 5 495 79
AY079315.1 - 4 - 5 421 79

View Full Product Details