Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001338928.1 | NP_001319661.1 | 11 - 12 | 3128 | 120 |
NM_001338929.1 | NP_001325749.1 | 11 - 12 | 3128 | 120 | |
NM_001338930.1 | NP_001325748.1 | 10 - 11 | 3132 | 120 | |
NM_001338931.1 | NP_001325750.1 | 8 - 9 | 2582 | 120 |