Assay Design
Gene Transcripts
- Gene Symbol
- ABCB17
- Entrez Gene ID
-
822467
- Gene Name
- P-glycoprotein 17
- Gene Aliases
-
AT3G28380, ATP-binding cassette B17, P-glycoprotein 17, PGP17
- Chromosome Location
-
Chr.3: 10623699 - 10628201 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_113758.2 |
NP_189479.1 |
1 - 2 |
241 |
86 |
View Full Product Details