Assay Design



Gene Transcripts

Gene Symbol
PHOT1
Entrez Gene ID
823721
Gene Name
phototropin 1
Gene Aliases
AT3G45780, F16L2.3, JK224, NONPHOTOTROPIC HYPOCOTYL 1, NPH1, PHOTOTROPIN, ROOT PHOTOTROPISM 1, RPT1, phototropin 1
Chromosome Location
Chr.3: 16816721 - 16824210 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035737.2 NP_001030814.1 6 - 7 1747 86
NM_114447.4 NP_190164.1 7 - 8 1745 86
GenBank mRNA AF030864.1 - 7 - 8 1600 86
AF360218.1 - 6 - 7 1554 86
AY040062.1 - 6 - 7 1537 86

View Full Product Details