Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001035737.2 | NP_001030814.1 | 8 - 9 | 1899 | 94 |
NM_114447.4 | NP_190164.1 | 9 - 10 | 1897 | 94 | |
GenBank mRNA | AF030864.1 | - | 9 - 10 | 1752 | 94 |
AF360218.1 | - | 8 - 9 | 1706 | 94 | |
AY040062.1 | - | 8 - 9 | 1689 | 94 |