Assay Design



Gene Transcripts

Gene Symbol
RUS1
Entrez Gene ID
823732
Gene Name
root UVB sensitive-like protein (Protein of unknown function, DUF647)
Gene Aliases
AT3G45890, ROOT UVB SENSITIVE 1, WEAK AUXIN RESPONSE 3, WXR3
Chromosome Location
Chr.3: 16871578 - 16874367 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_114458.4 NP_190175.2 2 - 3 1095 70
GenBank mRNA AK229694.1 - 2 - 3 992 70
BT008583.1 - 2 - 3 976 70
BT008683.1 - 2 - 3 992 70

View Full Product Details