Assay Design



Gene Transcripts

Gene Symbol
EB1a
Entrez Gene ID
823923
Gene Name
microtubule end binding protein EB1A
Gene Aliases
ARABIDOPSIS THALIANA MICROTUBULE END BINDING PROTEIN EB1A, AT3G47690, ATEB1-HOMOLOG2, ATEB1A, ATEB1H2, AtEB1a, microtubule end binding protein EB1A
Chromosome Location
Chr.3: 17581096 - 17583586 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_114637.4 NP_190353.3 4 - 5 626 101
GenBank mRNA AK227867.1 - 4 - 5 591 101
BT009707.1 - 4 - 5 369 101

View Full Product Details