Assay Design



Gene Transcripts

Gene Symbol
ABCA2
Entrez Gene ID
823927
Gene Name
ATP-binding cassette A2
Gene Aliases
A. THALIANA ABC2 HOMOLOG 1, ABC2 homolog 1, AT3G47730, ATATH1, ATH1, ATP-binding cassette A2
Chromosome Location
Chr.3: 17594060 - 17599083 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_114641.4 NP_190357.2 6 - 7 1504 66
GenBank mRNA BT004197.1 - 6 - 7 1317 66
BT005456.1 - 6 - 7 1249 66

View Full Product Details