Assay Design



Gene Transcripts

Gene Symbol
ABCA5
Entrez Gene ID
823930
Gene Name
ABC2 homolog 4
Gene Aliases
A. THALIANA ABC2 HOMOLOG 4, ABC2 homolog 4, AT3G47760, ATATH4, ATH4, ATP-binding cassette A5
Chromosome Location
Chr.3: 17611787 - 17616743 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001339333.1 NP_001326003.1 10 - 11 1371 68

View Full Product Details