Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_114669.4 | NP_190383.1 | 3 - 4 | 681 | 117 |
GenBank mRNA | AB030820.1 | - | 3 - 4 | 437 | 117 |
AF327426.1 | - | 3 - 4 | 431 | 117 | |
AF349447.1 | - | 3 - 4 | 437 | 117 | |
AF349522.1 | - | 3 - 4 | 355 | 117 | |
AF372911.1 | - | 3 - 4 | 431 | 117 | |
AY090443.1 | - | 3 - 4 | 435 | 117 | |
BT000752.1 | - | 3 - 4 | 380 | 117 | |
BT006371.1 | - | 3 - 4 | 355 | 117 |