Assay Design



Gene Transcripts

Gene Symbol
EDS1
Entrez Gene ID
823964
Gene Name
alpha/beta-Hydrolases superfamily protein
Gene Aliases
AT3G48090, ATEDS1, EDS1 PROTEIN, enhanced disease susceptibility 1
Chromosome Location
Chr.3: 17755329 - 17758147 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035752.1 NP_001030829.1 2 - 3 1035 113
NM_114678.4 NP_190392.1 2 - 3 1505 113
GenBank mRNA AK316830.1 - 2 - 3 1036 113
AY046025.1 - 2 - 3 1094 113
AY150423.1 - 2 - 3 1050 113

View Full Product Details