Assay Design



Gene Transcripts

Gene Symbol
RR5
Entrez Gene ID
823965
Gene Name
response regulator 5
Gene Aliases
ARABIDOPSIS THALIANA RESPONSE REGULATOR 2, ARR5, AT3G48100, ATRR2, IBC6, INDUCED BY CYTOKININ 6, response regulator 5
Chromosome Location
Chr.3: 17758427 - 17760950 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_114679.4 NP_190393.1 4 - 5 582 120
GenBank mRNA AB010916.1 - 4 - 5 506 120
AK226519.1 - 4 - 5 444 120
AY072390.1 - 4 - 5 441 120
AY114698.1 - 4 - 5 372 120

View Full Product Details