Assay Design
Gene Transcripts
- Gene Symbol
- MYB45
- Entrez Gene ID
-
824053
- Gene Name
- myb domain protein 45
- Gene Aliases
-
AT3G48920, AtMYB45, myb domain protein 45
- Chromosome Location
-
Chr.3: 18139172 - 18140772 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_114751.2 |
NP_190461.1 |
1 - 2 |
373 |
157 |
GenBank mRNA |
AY519596.1 |
- |
1 - 2 |
153 |
157 |
View Full Product Details