Assay Design
Gene Transcripts
- Gene Symbol
- iqd15
- Entrez Gene ID
-
824100
- Gene Name
- IQ-domain 15
- Gene Aliases
-
AT3G49380, IQ-domain 15
- Chromosome Location
-
Chr.3: 18311760 - 18313621 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_114798.2 |
NP_190507.1 |
1 - 2 |
276 |
129 |
View Full Product Details