Assay Design



Gene Transcripts

Gene Symbol
CEN2
Entrez Gene ID
824198
Gene Name
centrin2
Gene Aliases
AT3G50360, ATCEN2, CEN1, CENTRIN 1, centrin2
Chromosome Location
Chr.3: 18674137 - 18675761 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_114896.5 NP_190605.1 3 - 4 473 77
GenBank mRNA AJ009672.1 - 3 - 4 255 77
AK175359.1 - 3 - 4 244 77
AK175375.1 - 3 - 4 212 77
AK176828.1 - 3 - 4 212 77
AK176882.1 - 3 - 4 263 77
BT025522.1 - 2 - 3 189 77

View Full Product Details