Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001339479.1 | NP_001327730.1 | 3 - 4 | 931 | 108 |
NM_114926.4 | NP_190635.1 | 3 - 4 | 931 | 108 | |
GenBank mRNA | AF412114.1 | - | 3 - 4 | 722 | 108 |
AY090266.1 | - | 3 - 4 | 694 | 108 |