Assay Design
Gene Transcripts
- Gene Symbol
- BCS1
- Entrez Gene ID
-
824257
- Gene Name
- cytochrome BC1 synthesi
- Gene Aliases
-
AT3G50930, AtBCS1, AtOM66, OM66, Outer Mitochondrial membrane protein of 66 kDa, cytochrome BC1 synthesis
- Chromosome Location
-
Chr.3: 18929526 - 18932181 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_114953.3 |
NP_190662.2 |
1 - 1 |
2055 |
148 |
GenBank mRNA |
AY064981.1 |
- |
1 - 1 |
2055 |
148 |
View Full Product Details