Assay Design
Gene Transcripts
- Gene Symbol
- NDPK1
- Entrez Gene ID
-
826515
- Gene Name
- nucleoside diphosphate kinase
- Gene Aliases
-
AT4G09320, ATNDK1, NDK1, T30A10.80, T30A10_80, nucleoside diphosphate kinase 1
- Chromosome Location
-
Chr.4: 5923484 - 5924366 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_117000.3 |
NP_567346.2 |
2 - 3 |
103 |
119 |
GenBank mRNA |
AF017641.1 |
- |
2 - 3 |
103 |
119 |
View Full Product Details