Assay Design



Gene Transcripts

Gene Symbol
ATSPS4F
Entrez Gene ID
826603
Gene Name
Sucrose-phosphate synthase family protein
Gene Aliases
AT4G10120, F28M11.40, F28M11_40
Chromosome Location
Chr.4: 6313914 - 6320094 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001036532.3 NP_001031609.1 4 - 5 784 119
NM_001340652.1 NP_001329003.1 3 - 4 1725 119
NM_001340653.1 NP_001329002.1 2 - 3 1013 119
NM_117080.5 NP_192750.2 4 - 5 1634 119
GenBank mRNA KJ138763.1 - 3 - 4 606 119

View Full Product Details