Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001084903.1 | NP_001078372.1 | 2 - 3 | 496 | 128 |
NM_117160.4 | NP_192830.1 | 2 - 3 | 563 | 128 | |
GenBank mRNA | AF053303.1 | - | 2 - 3 | 444 | 128 |
AK228302.1 | - | 2 - 3 | 421 | 128 | |
AK229220.1 | - | 2 - 3 | 511 | 128 | |
AY085896.1 | - | 2 - 3 | 490 | 128 | |
BT005847.1 | - | 2 - 3 | 400 | 128 |