Assay Design



Gene Transcripts

Gene Symbol
NDPK3
Entrez Gene ID
826702
Gene Name
nucleoside diphosphate kinase 3
Gene Aliases
AT4G11010, T22B4.4, nucleoside diphosphate kinase 3
Chromosome Location
Chr.4: 6732420 - 6734454 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_117171.4 NP_192839.1 1 - 2 241 136
GenBank mRNA AF044265.1 - 1 - 2 115 136
AY035184.1 - 1 - 2 148 136
AY062964.1 - 1 - 2 85 136

View Full Product Details