Assay Design



Gene Transcripts

Gene Symbol
EDR2
Entrez Gene ID
827642
Gene Name
ENHANCED DISEASE RESISTANCE 2
Gene Aliases
AT4G19040, ENHANCED DISEASE RESISTANCE 2, F13C5.210, F13C5_210
Chromosome Location
Chr.4: 10431515 - 10437564 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001203840.1 NP_001190769.1 14 - 15 1634 91
NM_001341322.1 NP_001319993.1 14 - 15 1776 91
NM_118022.5 NP_193639.2 14 - 15 1776 91

View Full Product Details