Assay Design
Gene Transcripts
- Gene Symbol
- EDR2
- Entrez Gene ID
-
827642
- Gene Name
- ENHANCED DISEASE RESISTANCE 2
- Gene Aliases
-
AT4G19040, ENHANCED DISEASE RESISTANCE 2, F13C5.210, F13C5_210
- Chromosome Location
-
Chr.4: 10431515 - 10437564 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_118022.5 |
NP_193639.2 |
15 - 16 |
1820 |
71 |
View Full Product Details