Assay Design
Gene Transcripts
- Gene Symbol
- INDH
- Entrez Gene ID
-
827696
- Gene Name
- INDH1(iron-sulfur protein required for NADH dehydrogenase)
- Gene Aliases
-
AT4G19540, F24J7.100, F24J7_100, IND1(iron-sulfur protein required for NADH dehydrogenase)-like, INDL, iron-sulfur protein required for NADH dehydrogenase
- Chromosome Location
-
Chr.4: 10657289 - 10659566 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_118074.2 |
NP_193689.1 |
3 - 4 |
538 |
133 |
GenBank mRNA |
AK229020.1 |
- |
3 - 4 |
368 |
133 |
View Full Product Details