Assay Design



Gene Transcripts

Gene Symbol
HSP21
Entrez Gene ID
828881
Gene Name
heat shock protein 21
Gene Aliases
AT4G27670, T29A15.160, T29A15_160, heat shock protein 21
Chromosome Location
Chr.4: 13818825 - 13819971 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_118906.3 NP_194497.1 1 - 2 325 70
GenBank mRNA DQ446875.1 - 1 - 2 249 70
X54102.1 - - 325 70

View Full Product Details