Assay Design



Gene Transcripts

Gene Symbol
VPS26B
Entrez Gene ID
828883
Gene Name
vacuolar protein sorting 26B
Gene Aliases
AT4G27690, T29A15.180, T29A15_180, vacuolar protein sorting 26B
Chromosome Location
Chr.4: 13823510 - 13826395 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001036655.1 NP_001031732.1 7 - 8 520 73
NM_118908.5 NP_194499.2 9 - 10 708 73
GenBank mRNA AY094409.1 - 9 - 10 621 73
AY122891.1 - 7 - 8 494 73

View Full Product Details