Assay Design



Gene Transcripts

Gene Symbol
ULT1
Entrez Gene ID
828934
Gene Name
Developmental regulator, ULTRAPETALA
Gene Aliases
AT4G28190, F26K10.70, F26K10_70, ULT, ULTRAPETALA, ULTRAPETALA1
Chromosome Location
Chr.4: 13984695 - 13987510 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_118959.5 NP_567799.1 1 - 2 1273 95
GenBank mRNA AK117106.1 - 1 - 2 310 95
BT026413.1 - 1 - 2 215 95

View Full Product Details