Assay Design
Gene Transcripts
- Gene Symbol
- RPN1B
- Entrez Gene ID
-
828964
- Gene Name
- 26S proteasome regulatory subunit S2 1B
- Gene Aliases
-
26S proteasome regulatory subunit S2 1B, AT4G28470, ATRPN1B, F20O9.150, F20O9_150
- Chromosome Location
-
Chr.4: 14066731 - 14072582 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_118989.6 |
NP_194576.5 |
17 - 18 |
2275 |
71 |
GenBank mRNA |
AY230829.1 |
- |
17 - 18 |
2109 |
71 |
View Full Product Details