Assay Design



Gene Transcripts

Gene Symbol
PHT4;5
Entrez Gene ID
832160
Gene Name
phosphate transporter 4;5
Gene Aliases
AT5G20380, F5O24.270, F5O24_270, phosphate transporter 4;5
Chromosome Location
Chr.5: 6887636 - 6892571 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001343659.1 NP_001330368.1 9 - 10 1249 121
NM_122045.4 NP_197538.2 9 - 10 1137 121

View Full Product Details