Assay Design



Gene Transcripts

Gene Symbol
MGD2
Entrez Gene ID
832163
Gene Name
monogalactosyldiacylglycerol synthase 2
Gene Aliases
ARABIDOPSIS THALIANA MONOGALACTOSYLDIACYLGLYCEROL SYNTHASE 2, AT5G20410, ATMGD2, F5O24.300, F5O24_300, monogalactosyldiacylglycerol synthase 2
Chromosome Location
Chr.5: 6896463 - 6898783 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_122048.4 NP_568394.2 3 - 4 1307 75
GenBank mRNA AJ000331.1 - 3 - 4 1093 75
KJ138675.1 - 3 - 4 1005 75

View Full Product Details