Assay Design



Gene Transcripts

Gene Symbol
RAD51
Entrez Gene ID
832208
Gene Name
RAS associated with diabetes protein 51
Gene Aliases
AT5G20850, ATRAD51, F22D1.20, F22D1_20, RAS associated with diabetes protein 51
Chromosome Location
Chr.5: 7070491 - 7072968 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001343688.1 NP_001332342.1 3 - 4 458 79
NM_122092.3 NP_568402.1 3 - 4 456 79

View Full Product Details