Assay Design



Gene Transcripts

Gene Symbol
RAD51
Entrez Gene ID
832208
Gene Name
RAS associated with diabetes protein 51
Gene Aliases
AT5G20850, ATRAD51, F22D1.20, F22D1_20, RAS associated with diabetes protein 51
Chromosome Location
Chr.5: 7070491 - 7072968 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001343688.1 NP_001332342.1 7 - 8 892 89
NM_122092.3 NP_568402.1 7 - 8 890 89
GenBank mRNA U43528.1 - 7 - 8 834 89

View Full Product Details