Assay Design



Gene Transcripts

Gene Symbol
ELM1
Entrez Gene ID
832295
Gene Name
fission ELM1-like protein (DUF1022)
Gene Aliases
AT5G22350, ELONGATED MITOCHONDRIA 1, MWD9.14, MWD9_14
Chromosome Location
Chr.5: 7397386 - 7401290 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_122140.4 NP_568417.1 2 - 3 976 63
GenBank mRNA AB379589.1 - 2 - 3 432 63
AY059915.1 - 2 - 3 624 63
AY128808.1 - 2 - 3 432 63

View Full Product Details