Assay Design



Gene Transcripts

Gene Symbol
ELM1
Entrez Gene ID
832295
Gene Name
fission ELM1-like protein (DUF1022)
Gene Aliases
AT5G22350, ELONGATED MITOCHONDRIA 1, MWD9.14, MWD9_14
Chromosome Location
Chr.5: 7397386 - 7401290 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_122140.4 NP_568417.1 8 - 9 1621 62
GenBank mRNA AB379589.1 - 8 - 9 1077 62
AY059915.1 - 8 - 9 1269 62
AY128808.1 - 8 - 9 1077 62

View Full Product Details