Assay Design



Gene Transcripts

Gene Symbol
FAR7
Entrez Gene ID
832303
Gene Name
fatty acid reductase 7
Gene Aliases
AT5G22420, MWD9.22, MWD9_22, fatty acid reductase 7
Chromosome Location
Chr.5: 7429769 - 7432825 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001343726.1 NP_001331820.1 5 - 6 1496 78
NM_001343727.1 NP_001331821.1 6 - 7 1167 78
NM_001343728.1 NP_001331818.1 7 - 8 1060 78
NM_001343729.1 NP_001331819.1 6 - 7 959 78
NM_001343730.1 NP_001331816.1 5 - 6 1014 78
NM_001343731.1 NP_001331817.1 3 - 4 541 78
NM_122147.2 NP_197634.1 6 - 7 708 78

View Full Product Details