Assay Design
Gene Transcripts
- Gene Symbol
- RAD5
- Entrez Gene ID
-
832338
- Gene Name
- DNA/RNA helicase protein
- Gene Aliases
-
AT5G22750, MDJ22.17, MDJ22_17, RAD5A
- Chromosome Location
-
Chr.5: 7565037 - 7571109 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_122181.3 |
NP_197667.1 |
3 - 4 |
1047 |
80 |
GenBank mRNA |
AK228695.1 |
- |
3 - 4 |
1021 |
80 |
View Full Product Details