Assay Design



Gene Transcripts

Gene Symbol
MAM1
Entrez Gene ID
832365
Gene Name
methylthioalkylmalate synthase 1
Gene Aliases
2-ISOPROPYLMALATE SYNTHASE, 2-ISOPROPYLMALATE SYNTHASE 3, AT5G23010, IMS3, METHYLTHIOALKYLMALATE SYNTHASE, T20O7.3, T20O7_3, methylthioalkylmalate synthase 1
Chromosome Location
Chr.5: 7703041 - 7706973 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001343784.1 NP_001330047.1 1 - 2 574 70
NM_001343785.1 NP_001330048.1 1 - 2 523 70
NM_122207.3 NP_197692.1 1 - 2 523 70
GenBank mRNA AJ486884.1 - 1 - 2 442 70
AJ486885.1 - 1 - 2 442 70
AJ486886.1 - 1 - 2 442 70
AJ486887.1 - 1 - 2 442 70
AY049037.1 - 1 - 2 442 70
AY054203.1 - 1 - 2 514 70
AY070471.1 - 1 - 2 520 70
AY149926.1 - 1 - 2 442 70

View Full Product Details