Assay Design



Gene Transcripts

Gene Symbol
AAE17
Entrez Gene ID
832369
Gene Name
acyl-activating enzyme 17
Gene Aliases
AT5G23050, MYJ24.4, MYJ24_4, acyl-activating enzyme 17
Chromosome Location
Chr.5: 7731221 - 7735566 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001343787.1 NP_001332667.1 7 - 8 1245 65
NM_122211.3 NP_197696.2 7 - 8 1245 65

View Full Product Details