Assay Design



Gene Transcripts

Gene Symbol
FSD3
Entrez Gene ID
832395
Gene Name
Fe superoxide dismutase 3
Gene Aliases
AT5G23310, Fe superoxide dismutase 3, MKD15.17, MKD15_17, SOD3, superoxide dismutase 3
Chromosome Location
Chr.5: 7850444 - 7852534 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_122237.4 NP_197722.1 6 - 7 822 74
GenBank mRNA AY065458.1 - 6 - 7 718 74
AY091225.1 - 6 - 7 642 74

View Full Product Details