Assay Design



Gene Transcripts

Gene Symbol
CDPK9
Entrez Gene ID
832424
Gene Name
calmodulin-like domain protein kinase 9
Gene Aliases
ARABIDOPSIS THALIANA CALMODULIN-LIKE DOMAIN PROTEIN KINASE 9, AT5G23580, ATCDPK9, ATCPK12, CALCIUM-DEPENDENT PROTEIN KINASE, CALCIUM-DEPENDENT PROTEIN KINASE 12, CPK12, calmodulin-like domain protein kinase 9
Chromosome Location
Chr.5: 7950202 - 7952619 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_122264.5 NP_197748.1 1 - 2 713 63
GenBank mRNA AK229078.1 - 1 - 2 586 63
BT026382.1 - 1 - 2 527 63
U20388.1 - 1 - 2 564 63

View Full Product Details